Assay Details
| Gene Symbol: Mvb12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175184.4 | NP_780393.2 | 7 - 8 | 1306 | 89 | |
| GenBank mRNA | AK012178.1 | - | 7 - 8 | 799 | 89 | |
| AK087821.1 | - | 7 - 8 | 824 | 89 | ||
| AK131113.1 | - | 7 - 8 | 824 | 89 | ||
| AK154267.1 | - | 7 - 8 | 801 | 89 | ||
| BC049129.1 | - | 7 - 8 | 819 | 6825067 | 89 | |
| BC059907.1 | - | 7 - 8 | 1306 | 6824889 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mvb12b
Genomic Map