Assay Details
| Gene Symbol: Lin9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103182.2 | NP_001096652.2 | 11 - 12 | 1335 | 141 | |
| NR_120498.1 | 11 - 12 | 1333 | 141 | |||
| GenBank mRNA | AF190325.1 | - | 11 - 12 | 1151 | 141 | |
| AK052618.1 | - | 11 - 12 | 1133 | 141 | ||
| AK089989.1 | - | 11 - 12 | 1337 | 141 | ||
| BC069181.1 | - | 8 - 9 | 874 | 141 | ||
| BC088992.1 | - | 11 - 12 | 1123 | 141 | ||
More Information
Set Membership: |
|

Lin9
Genomic Map