Assay Details
| Gene Symbol: Lin9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103182.2 | NP_001096652.2 | 3 - 4 | 381 | 113 | |
| NR_120498.1 | 3 - 4 | 381 | 113 | |||
| GenBank mRNA | AF190325.1 | - | 3 - 4 | 197 | 113 | |
| AK042357.1 | - | 3 - 4 | 163 | 113 | ||
| AK052618.1 | - | 3 - 4 | 179 | 113 | ||
| AK089989.1 | - | 3 - 4 | 382 | 113 | ||
| BC088992.1 | - | 3 - 4 | 171 | 113 | ||
| BC099695.1 | - | 3 - 4 | 165 | 113 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF190325.1, AK042357.1 transcripts.
Set Membership: |
|

Lin9
Genomic Map