Assay Details
| Gene Symbol: Lin9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103182.2 | NP_001096652.2 | 8 - 9 | 1035 | 94 | |
| GenBank mRNA | AF190325.1 | - | 8 - 9 | 851 | 94 | |
| AK042357.1 | - | 8 - 9 | 791 | 94 | ||
| AK052618.1 | - | 8 - 9 | 833 | 94 | ||
| AK089989.1 | - | 8 - 9 | 1037 | 94 | ||
| BC099695.1 | - | 8 - 9 | 819 | 94 | ||
More Information
Set Membership: |
|

Lin9
Genomic Map