Assay Details
| Gene Symbol: Lin9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103182.2 | NP_001096652.2 | 9 - 10 | 1155 | 92 | |
| NR_120498.1 | 9 - 10 | 1153 | 92 | |||
| GenBank mRNA | AF190325.1 | - | 9 - 10 | 971 | 92 | |
| AK042357.1 | - | 9 - 10 | 911 | 92 | ||
| AK052618.1 | - | 9 - 10 | 953 | 92 | ||
| AK089989.1 | - | 9 - 10 | 1157 | 92 | ||
| BC069181.1 | - | 6 - 7 | 694 | 92 | ||
| BC088992.1 | - | 9 - 10 | 943 | 92 | ||
| BC099695.1 | - | 9 - 10 | 939 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lin9
Genomic Map