Assay Details
| Gene Symbol: Sppl2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175195.3 | NP_780404.2 | 11 - 12 | 1225 | 58 | |
| GenBank mRNA | AK046804.1 | - | 11 - 12 | 1244 | 58 | |
| AK148109.1 | - | 11 - 12 | 1192 | 58 | ||
| AK157906.1 | - | 11 - 12 | 1170 | 58 | ||
| AK166364.1 | - | 11 - 12 | 1176 | 58 | ||
| AK170375.1 | - | 11 - 12 | 1191 | 58 | ||
| AK194712.1 | - | 1 - 2 | 35 | 58 | ||
| BC034206.1 | - | 8 - 9 | 780 | 58 | ||
| BC052094.1 | - | 9 - 10 | 899 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sppl2b
Genomic Map