Assay Details
| Gene Symbol: Srrm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175229.3 | NP_780438.2 | 5 - 6 | 620 | 70 | |
| GenBank mRNA | AK090123.1 | - | 5 - 6 | 581 | 70 | |
| AK171001.1 | - | 6 - 7 | 893 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srrm2
Genomic Map