Assay Details
| Gene Symbol: Adhfe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175236.4 | NP_780445.1 | 6 - 7 | 593 | 77 | |
| NR_027664.1 | 5 - 6 | 552 | 77 | |||
| GenBank mRNA | AK018241.1 | - | 6 - 7 | 567 | 77 | |
| AK038853.1 | - | 6 - 7 | 560 | 77 | ||
| AK046780.1 | - | 6 - 7 | 519 | 77 | ||
| AK050178.1 | - | 5 - 6 | 517 | 77 | ||
| AK084104.1 | - | 521 | 77 | |||
| BC026584.1 | - | 6 - 7 | 551 | 4972323 | 77 | |
| CA467848.1 | - | 6 - 7 | 632 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adhfe1
Genomic Map