Assay Details
| Gene Symbol: Adhfe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175236.4 | NP_780445.1 | 8 - 9 | 769 | 67 | |
| NR_027664.1 | 7 - 8 | 728 | 67 | |||
| GenBank mRNA | AK038853.1 | - | 8 - 9 | 736 | 67 | |
| AK046780.1 | - | 8 - 9 | 695 | 67 | ||
| AK050178.1 | - | 7 - 8 | 693 | 67 | ||
| AK084104.1 | - | 697 | 67 | |||
| BC026584.1 | - | 8 - 9 | 727 | 4972323 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adhfe1
Genomic Map