Assay Details
| Gene Symbol: Sec24a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290785.1 | NP_001277714.1 | 20 - 21 | 3620 | 66 | |
| NM_175255.3 | NP_780464.2 | 20 - 21 | 3623 | 66 | ||
| GenBank mRNA | AK029139.1 | - | 20 - 21 | 3376 | 66 | |
| AK155182.1 | - | 20 - 21 | 3409 | 66 | ||
| BC117905.1 | - | 20 - 21 | 2887 | 66 | ||
| BG065014.2 | - | 3 - 4 | 66 | |||
| CJ055291.1 | - | 3 - 4 | 283 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec24a
Genomic Map