Assay Details
| Gene Symbol: Epm2aip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK030084.1 | - | 1 - 2 | 2595 | 150 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26828616 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26828614 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26828615 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P482043 | AK122366.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P482043 | CN530473.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P482043 | NM_175266.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P482043 | AK030084.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P482043 | AK122366.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P482043 | CN530473.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P482043 | NM_175266.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P482043 | AK030084.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124676 | AK033908.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124676 | AK043518.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124676 | AK032565.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124676 | NM_175266.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 446370 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 327784 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 966529 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 136743 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 180397 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 96025 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113267 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124676 | AK033908.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124676 | AK043518.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124676 | AK032565.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124676 | NM_175266.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P482043 | AK122366.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P482043 | CN530473.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P482043 | NM_175266.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P482043 | AK030084.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P482043 | AK122366.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P482043 | CN530473.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P482043 | NM_175266.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P482043 | AK030084.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P482043 | AK122366.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P482043 | CN530473.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P482043 | NM_175266.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P482043 | AK030084.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P482043 | AK122366.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P482043 | CN530473.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P482043 | NM_175266.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P482043 | AK030084.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828641 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828631 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828646 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828615 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26828624 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113267 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06927432 | - | - |
| Whole Mouse Genome G4122A | A_51_P482043 | AK122366.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P482043 | CN530473.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P482043 | NM_175266.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P482043 | AK030084.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P482043 | AK122366.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P482043 | CN530473.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P482043 | NM_175266.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P482043 | AK030084.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P37522 | AK043518.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P37522 | AK122366.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P37522 | AK032284.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P37522 | NM_175266.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P37522 | AK030084.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108906 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113267 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08040483 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08040469 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08040463 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26828616 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26828614 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26828615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26828639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07600405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08040468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07600400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08040486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08040505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26828691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07505334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07463984 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10589759 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589757 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589760 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589758 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434106_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454835_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434105_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425624_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10589756 | - | - |
| Mouse Genome 430 2.0 Array | 1425624_at | - | - |
| Mouse Genome 430 2.0 Array | 1454835_at | - | - |
| Mouse Genome 430 2.0 Array | 1434105_at | - | - |
| Mouse Genome 430 2.0 Array | 1425624_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454835_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434105_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434106_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434106_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 77781_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 77781_PM_at | - | - |
| Mouse Expression Set 430 | 1425624_PM_at | - | - |
| Mouse Expression Set 430 | 1425624_at | - | - |
| Mouse Expression Set 430 | 1434106_PM_at | - | - |
| Mouse Expression Set 430 | 1434105_PM_at | - | - |
| Mouse Expression Set 430 | 1434105_at | - | - |
| Mouse Expression Set 430 | 1454835_PM_at | - | - |
| Mouse Expression Set 430 | 1454835_at | - | - |
| Mouse Expression Set 430 | 1434106_at | - | - |
| Murine Genome U74v2 Set | 129161_at | - | - |
| Murine Genome U74v2 Set | 129161_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425624_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425624_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425624_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425624_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454835_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434105_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434106_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434106_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454835_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434105_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3360452 | - | - |
| MouseWG-6 v2 Expression BeadChip | 6590736 | - | - |
| MouseWG-6 v2 Expression BeadChip | 102450156 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6590736 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100540446 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3360452 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102450156 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006590736 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003360452 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2699995 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1256740 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221110 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2699995 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226367 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1256740 | - | - |
| mouse-6 v1.1 (Illumina) | scl077781.2_11-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0077781.2_58-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100540446 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3360452 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6590736 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102450156 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0077781.2_58-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl077781.2_11-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6590736 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102450156 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100540446 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3360452 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1256740 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2699995 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2895177 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2895177 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1256740 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2699995 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221110 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1226367 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0077781.2_58-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl077781.2_11-S | - | - |

Epm2aip1
Genomic Map