Assay Details
| Gene Symbol: Rtfdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK030734.1 | - | 1 | 3447 | 103 | |
| Gene Symbol: Gcnt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039560.3 | NP_001034649.1 | 4 | 103 | ||
| GenBank mRNA | AK039414.1 | - | 4 | 103 | ||
| AK132647.1 | - | 2 | 103 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rtfdc1
Genomic Map