Assay Details
| Gene Symbol: Plekhg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081333.1 | NP_001074802.1 | 14 - 15 | 2513 | 106 | |
| GenBank mRNA | AK029859.1 | - | 12 - 13 | 3185 | 106 | |
| AK131125.1 | - | 10 - 11 | 3480 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plekhg4
Genomic Map