Assay Details
| Gene Symbol: Plekhg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081333.1 | NP_001074802.1 | 17 - 18 | 3084 | 114 | |
| GenBank mRNA | AK029859.1 | - | 15 - 16 | 3756 | 114 | |
| AK131125.1 | - | 11 - 12 | 4215 | 114 | ||
| BC024792.1 | - | 1 - 2 | 537 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plekhg4
Genomic Map