Assay Details
| Gene Symbol: Plekhg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081333.1 | NP_001074802.1 | 18 - 19 | 3206 | 103 | |
| GenBank mRNA | AK029859.1 | - | 16 - 17 | 3878 | 103 | |
| AK131125.1 | - | 12 - 13 | 4337 | 103 | ||
| BC024792.1 | - | 2 - 3 | 659 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plekhg4
Genomic Map