Assay Details
| Gene Symbol: Plekhg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081333.1 | NP_001074802.1 | 5 - 6 | 866 | 78 | |
| GenBank mRNA | AK029859.1 | - | 6 - 7 | 862 | 78 | |
| AK131125.1 | - | 4 - 5 | 973 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029859.1 transcripts.
Set Membership: |
|

Plekhg4
Genomic Map