Assay Details
| Gene Symbol: Cacng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK047238.1 | - | 1252 | 100 | ||
| AK164203.1 | - | 1337 | 100 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cacng2
Genomic Map