Assay Details
| Gene Symbol: Ttbk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162864.1 | NP_001156336.1 | 15 | 6794 | 79 | |
| GenBank mRNA | AK049158.1 | - | 1 | 1756 | 79 | |
| AK173279.1 | - | 2 | 3758 | 79 | ||
| BC059249.1 | - | 2 | 3998 | 79 | ||
| BC094327.1 | - | 1 | 2079 | 79 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
biological_process
regulation of cell shape
phosphorylation
peptidyl-serine phosphorylation
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein phosphorylation
biological_process
regulation of cell shape
phosphorylation
peptidyl-serine phosphorylation
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31206982 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31206981 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31206983 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31206980 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2082215 | AK173279.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2082215 | BC059249.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2082215 | BC094327.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2082215 | NM_001162864.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2082215 | AK049158.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124070 | AK173279.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124070 | BC059249.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124070 | NM_001162864.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 878921 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930177 | AK132617.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930177 | AK149439.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930177 | BC010761.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930177 | BC011233.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930177 | BC046605.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930177 | BC058743.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930177 | BC063778.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930177 | J03549.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930177 | M19319.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930177 | NM_007812.4 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930177 | NM_009997.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930177 | X89864.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930177 | XM_001478723.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930177 | XM_286199.6 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 514667 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 154703 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 656031 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 295536 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 621022 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 794603 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 627344 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 370663 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 103640 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 872833 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655359 | NM_001042501.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 646715 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 802962 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 881474 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61110 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 450951 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 113783 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588638 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 9308 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736652 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 497790 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 389243 | AB028737.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 389243 | BC040400.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 389243 | BC049980.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 389243 | EF109740.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 389243 | EF109741.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 389243 | NM_018795.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 85313 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 917596 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416782 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 391809 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 662143 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 64693 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 164911 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 141134 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 149039 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 28226 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150622 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2082215 | AK173279.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2082215 | BC059249.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2082215 | BC094327.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2082215 | NM_001162864.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2082215 | AK049158.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124070 | AK173279.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124070 | BC059249.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124070 | NM_001162864.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P455077 | NM_001162864.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P455077 | AK144536.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P336293 | AK173279.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P336293 | BC059249.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P336293 | BC094327.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P336293 | NM_001162864.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P336293 | AK049158.1 | mouse_transcriptome |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31206982 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31206980 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31206981 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31206983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207158 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206984 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207156 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207173 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13504477 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13537639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207138 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13320010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207175 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207164 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13297801 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13260851 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13459468 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206965 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13537643 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207145 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206961 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207166 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207178 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207139 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13220684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13570831 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13260853 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13547643 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206972 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207171 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13159458 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207161 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207169 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13340515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13320015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13321883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207143 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13448446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13214566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31206985 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31207147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13321881 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10451306 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451310 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451321 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451312 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451311 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451307 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451318 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451309 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451320 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451316 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451317 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451314 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451305 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451308 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451313 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451319 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451304 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451315 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1443528_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10451303 | - | - |
| Mouse Genome 430 2.0 Array | 1443528_at | - | - |
| Mouse Genome 430 2.0 Array | 1443528_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 106763_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 106763_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700020457.mm.1 | NM_001162864.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020457.mm.1 | NM_001162864.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020457.mm.1 | TC1700002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700020461.mm.1 | NM_001162864.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020461.mm.1 | NM_001162864.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020461.mm.1 | TC1700002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700020466.mm.1 | NM_001162864.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020466.mm.1 | NM_001162864.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020466.mm.1 | TC1700002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700020460.mm.1 | NM_001162864.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020460.mm.1 | NM_001162864.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020460.mm.1 | TC1700002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700020469.mm.1 | NM_001162864.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020469.mm.1 | NM_001162864.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020469.mm.1 | TC1700002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700020459.mm.1 | NM_001162864.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020459.mm.1 | NM_001162864.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020459.mm.1 | TC1700002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700020471.mm.1 | NM_001162864.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020471.mm.1 | NM_001162864.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020471.mm.1 | TC1700002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700020462.mm.1 | NM_001162864.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020462.mm.1 | NM_001162864.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020462.mm.1 | TC1700002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700020463.mm.1 | NM_001162864.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020463.mm.1 | NM_001162864.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020463.mm.1 | TC1700002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700020467.mm.1 | NM_001162864.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020467.mm.1 | NM_001162864.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020467.mm.1 | TC1700002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700020456.mm.1 | NM_001162864.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020456.mm.1 | NM_001162864.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020456.mm.1 | TC1700002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700020470.mm.1 | NM_001162864.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020470.mm.1 | NM_001162864.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020470.mm.1 | TC1700002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700020465.mm.1 | NM_001162864.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020465.mm.1 | NM_001162864.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020465.mm.1 | TC1700002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700020464.mm.1 | NM_001162864.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020464.mm.1 | NM_001162864.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020464.mm.1 | TC1700002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700020468.mm.1 | NM_001162864.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020468.mm.1 | NM_001162864.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020468.mm.1 | TC1700002177.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1443528_PM_at | - | - |
| Mouse Expression Set 430 | 1445479_at | - | - |
| Mouse Expression Set 430 | 1443528_at | - | - |
| Mouse Expression Set 430 | 1445479_PM_at | - | - |
| Murine Genome U74v2 Set | 138020_at | - | - |
| Murine Genome U74v2 Set | 138020_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443528_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1443528_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 322042 | - | - |
| ABI Mouse Genome Survey Microarray | 352691 | - | - |

Ttbk1
Genomic Map