Assay Details
| Gene Symbol: Ccdc122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175369.3 | NP_780578.1 | 2 - 3 | 300 | 96 | |
| GenBank mRNA | AK077161.1 | - | 2 - 3 | 301 | 96 | |
| BC096577.1 | - | 2 - 3 | 294 | 96 | ||
| BC138881.1 | - | 2 - 3 | 208 | 8861903 | 96 | |
| BC138883.1 | - | 2 - 3 | 208 | 8861905 | 96 | |
| BC145455.1 | - | 2 - 3 | 208 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc122
Genomic Map