Assay Details
| Gene Symbol: Als2cr12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175370.4 | NP_780579.1 | 14 - 15 | 1263 | 102 | |
| GenBank mRNA | AK077190.1 | - | 14 - 15 | 1231 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2150876 | NM_175370.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2150876 | AI642686.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2150876 | AK077190.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 956271 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868158 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576338 | AK032023.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576338 | AK049830.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576338 | AY348866.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576338 | BC059811.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576338 | NM_172994.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 295113 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904420 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 616937 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 902846 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 747057 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 2424 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 556991 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 351419 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 526511 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 450094 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 365005 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 37499 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 113511 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 36853 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 79528 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113188 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143732 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2150876 | NM_175370.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2150876 | AI642686.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2150876 | AK077190.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20260141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20260107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20260137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20260118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20260111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20260135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20260128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20260102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20260136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20260116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20260127 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113188 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00077161 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143732 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00077139 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04110826 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04110891 | - | - |
| Whole Mouse Genome G4122A | A_52_P93380 | BC145040.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P93380 | AK133373.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P93380 | BC145039.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P93380 | NM_175370.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P93380 | AK016678.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102762 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117542 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113188 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143732 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01062750 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00931674 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01062751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20259995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20259998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20259991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20259993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20259997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20259950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20259978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20259923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20259972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20259933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20259940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20259942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20260135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20259933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20259963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20259915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20259961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20260127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20260113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00965144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20260116 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00810965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20259950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20260135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20259908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20260111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20260091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20260126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20260088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00502699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20260118 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00481624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01111884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20259942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20259940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20260121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20260107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20260077 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20259905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01064483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20260100 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20260106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20260136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20259945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20260094 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20260133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20259907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20260147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20259914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20259923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20260102 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20259949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20259925 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10354889 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354892 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354885 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354894 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354888 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354893 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354896 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354886 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354890 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354887 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354884 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354895 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10354891 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431882_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439758_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10354883 | - | - |
| Mouse Genome 430 2.0 Array | 1431882_at | - | - |
| Mouse Genome 430 2.0 Array | 1439758_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431882_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439758_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 108812_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 108812_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100019459.mm.1 | NM_175370.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100019459.mm.1 | TC0100002388.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100019457.mm.1 | NM_175370.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100019457.mm.1 | TC0100002388.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100019456.mm.1 | NM_175370.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100019456.mm.1 | TC0100002388.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1439758_PM_at | - | - |
| Mouse Expression Set 430 | 1431882_PM_at | - | - |
| Mouse Expression Set 430 | 1431882_at | - | - |
| Mouse Expression Set 430 | 1439758_at | - | - |
| Murine Genome U74v2 Set | 167658_PM_at | - | - |
| Murine Genome U74v2 Set | 167658_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439758_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431882_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431882_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439758_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 522214 | - | - |
| ABI Mouse Genome Survey Microarray | 747399 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101400400 | - | - |
| MouseWG-6 v2 Expression BeadChip | 6900204 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6900204 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101400400 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006900204 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2698213 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2698213 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1214562 | - | - |
| mouse-6 v1.1 (Illumina) | scl16728.15.1_237-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101400400 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6900204 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl16728.15.1_237-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101400400 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6900204 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2698213 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2698213 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1214562 | - | - |

Als2cr12
Genomic Map