Assay Details
| Gene Symbol: Mtrf1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175374.3 | NP_780583.1 | 2 - 3 | 492 | 92 | |
| GenBank mRNA | AK036424.1 | - | 2 - 3 | 396 | 92 | |
| AK078897.1 | - | 2 - 3 | 460 | 92 | ||
| AK185802.1 | - | 2 - 3 | 99 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtrf1l
Genomic Map