Assay Details
| Gene Symbol: Tspan9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175414.4 | NP_780623.1 | 6 - 7 | 614 | 72 | |
| GenBank mRNA | AK079139.1 | - | 6 - 7 | 587 | 72 | |
| AK158729.1 | - | 5 - 6 | 499 | 72 | ||
| AK163395.1 | - | 6 - 7 | 670 | 72 | ||
| BC052503.1 | - | 6 - 7 | 596 | 5146854 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tspan9
Genomic Map