Assay Details
| Gene Symbol: Aldh4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175438.4 | NP_780647.3 | 11 - 12 | 1430 | 64 | |
| GenBank mRNA | AK044712.1 | - | 11 - 12 | 1226 | 64 | |
| BC024133.1 | - | 10 - 11 | 1148 | 64 | ||
| BC026589.1 | - | 6 - 7 | 642 | 64 | ||
| BC039281.1 | - | 11 - 12 | 1214 | 4975491 | 64 | |
| BC056226.1 | - | 11 - 12 | 1314 | 5064715 | 64 | |
| BC060650.1 | - | 10 - 11 | 2820 | 30354228 | 64 | |
| CD354968.1 | - | 1 - 2 | 62 | 64 | ||
| CK332989.1 | - | 5 - 6 | 540 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aldh4a1
Genomic Map