Assay Details
| Gene Symbol: Hid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175454.2 | NP_780663.1 | 6 - 7 | 840 | 71 | |
| GenBank mRNA | AB087506.1 | - | 6 - 7 | 840 | 71 | |
| AK044395.1 | - | 6 - 7 | 818 | 71 | ||
| AK138882.1 | - | 6 - 7 | 844 | 71 | ||
| BC024617.1 | - | 6 - 7 | 840 | 4190480 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hid1
Genomic Map