Assay Details
| Gene Symbol: Nmnat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175460.3 | NP_780669.1 | 4 - 5 | 330 | 64 | |
| GenBank mRNA | AK083532.1 | - | 4 - 5 | 434 | 64 | |
| AK084041.1 | - | 4 - 5 | 440 | 64 | ||
| AK129155.1 | - | 4 - 5 | 416 | 64 | ||
| BC089007.1 | - | 4 - 5 | 330 | 30532596 | 64 | |
More Information
Set Membership: |
|

Nmnat2
Genomic Map