Assay Details
| Gene Symbol: Nmnat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175460.3 | NP_780669.1 | 9 - 10 | 762 | 77 | |
| GenBank mRNA | AK083532.1 | - | 9 - 10 | 866 | 77 | |
| AK084041.1 | - | 9 - 10 | 872 | 77 | ||
| BC089007.1 | - | 9 - 10 | 762 | 30532596 | 77 | |
More Information
Set Membership: |
|

Nmnat2
Genomic Map