Assay Details
| Gene Symbol: Fam78b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160262.1 | NP_001153734.1 | 2 - 3 | 1658 | 81 | |
| GenBank mRNA | AK047703.1 | - | 2 - 3 | 1496 | 81 | |
More Information
Set Membership: |
|

Fam78b
Genomic Map