Assay Details
| Gene Symbol: Rspo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040689.1 | NP_001035779.1 | 2 - 3 | 416 | 98 | |
| GenBank mRNA | AK020904.1 | - | 2 - 3 | 417 | 98 | |
| AK043484.1 | - | 2 - 3 | 417 | 99 | ||
| BC048707.1 | - | 2 - 3 | 390 | 6511472 | 98 | |
| BC116367.1 | - | 2 - 3 | 379 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rspo4
Genomic Map