Assay Details
| Gene Symbol: Arhgap25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037727.2 | NP_001032816.1 | 3 - 4 | 720 | 71 | |
| NM_001286610.1 | NP_001273539.1 | 2 - 3 | 520 | 71 | ||
| NM_175476.4 | NP_780685.2 | 3 - 4 | 391 | 71 | ||
| NR_104483.1 | 3 - 4 | 391 | 71 | |||
| GenBank mRNA | AK037710.1 | - | 3 - 4 | 453 | 71 | |
| AK048450.1 | - | 3 - 4 | 350 | 71 | ||
| AK137444.1 | - | 3 - 4 | 379 | 71 | ||
| AK137526.1 | - | 3 - 4 | 328 | 71 | ||
| AK159598.1 | - | 2 - 3 | 521 | 71 | ||
| AK159941.1 | - | 2 - 3 | 521 | 71 | ||
| AK169897.1 | - | 3 - 4 | 697 | 71 | ||
| AK172878.1 | - | 1 - 2 | 50 | 71 | ||
| BC138751.1 | - | 2 - 3 | 299 | 8861773 | 71 | |
| BC138752.1 | - | 2 - 3 | 299 | 8861774 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap25
Genomic Map