Assay Details
| Gene Symbol: Grik4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175481.5 | NP_780690.2 | 11 - 12 | 1369 | 57 | |
| GenBank mRNA | AK032029.1 | - | 11 - 12 | 1370 | 57 | |
| AK080582.1 | - | 11 - 12 | 1497 | 57 | ||
| BC118010.2 | - | 10 - 11 | 1284 | 40096690 | 57 | |
More Information
Set Membership: |
|

Grik4
Genomic Map