Assay Details
| Gene Symbol: Grik4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175481.5 | NP_780690.2 | 19 - 20 | 2591 | 137 | |
| GenBank mRNA | AK032029.1 | - | 19 - 20 | 2592 | 137 | |
| AK080582.1 | - | 19 - 20 | 2721 | 137 | ||
| BC118010.2 | - | 18 - 19 | 2506 | 40096690 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|

Grik4
Genomic Map