Assay Details
| Gene Symbol: Grik4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175481.5 | NP_780690.2 | 5 - 6 | 548 | 87 | |
| GenBank mRNA | AK032029.1 | - | 5 - 6 | 549 | 87 | |
| AK080582.1 | - | 5 - 6 | 676 | 87 | ||
| BC118010.2 | - | 4 - 5 | 463 | 40096690 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Grik4
Genomic Map