Assay Details
| Gene Symbol: Ppm1k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175523.4 | NP_780732.1 | 1 - 2 | 146 | 118 | |
| GenBank mRNA | AK044610.1 | - | 1 - 2 | 216 | 118 | |
| BC092238.1 | - | 1 - 2 | 146 | 30642218 | 118 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK044610.1, BC092238.1, NM_175523.4 transcripts.
Set Membership: |
|

Ppm1k
Genomic Map