Assay Details
| Gene Symbol: Wdfy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175546.4 | NP_780755.2 | 2 - 3 | 429 | 64 | |
| GenBank mRNA | AK086275.1 | - | 2 - 3 | 442 | 64 | |
| AK131958.1 | - | 2 - 3 | 428 | 64 | ||
| AK151817.1 | - | 2 - 3 | 385 | 64 | ||
| AK169499.1 | - | 2 - 3 | 406 | 64 | ||
| BC116632.1 | - | 2 - 3 | 375 | 40056912 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdfy2
Genomic Map