Assay Details
| Gene Symbol: Wdfy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175546.4 | NP_780755.2 | 7 - 8 | 951 | 56 | |
| GenBank mRNA | AK045072.1 | - | 6 - 7 | 682 | 56 | |
| AK086275.1 | - | 7 - 8 | 964 | 56 | ||
| AK131958.1 | - | 7 - 8 | 950 | 56 | ||
| AK151817.1 | - | 7 - 8 | 907 | 56 | ||
| BC116632.1 | - | 7 - 8 | 897 | 40056912 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdfy2
Genomic Map