Assay Details
| Gene Symbol: Nlrc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081280.1 | NP_001074749.1 | 9 - 10 | 2799 | 72 | |
| NM_175547.3 | NP_780756.1 | 6 - 7 | 722 | 72 | ||
| GenBank mRNA | AK079766.1 | - | 6 - 7 | 646 | 72 | |
| AK084199.1 | - | 6 - 7 | 636 | 72 | ||
| AK171971.1 | - | 8 - 9 | 3812 | 72 | ||
| BC130223.1 | - | 6 - 7 | 2172 | 72 | ||
| BC152543.1 | - | 6 - 7 | 2181 | 72 | ||
More Information
Set Membership: |
|

Nlrc3
Genomic Map