Assay Details
| Gene Symbol: Plch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113360.2 | NP_001106831.1 | 14 - 15 | 2178 | 106 | |
| NM_175556.4 | NP_780765.2 | 14 - 15 | 2178 | 106 | ||
| GenBank mRNA | AK044619.1 | - | 14 - 15 | 2178 | 106 | |
| AK129147.1 | - | 14 - 15 | 1938 | 106 | ||
| AY966876.1 | - | 14 - 15 | 1956 | 106 | ||
| BC040465.1 | - | 2 - 3 | 189 | 106 | ||
| BC052329.1 | - | 4 - 5 | 311 | 106 | ||
| DQ176851.1 | - | 14 - 15 | 2178 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plch2
Genomic Map