Assay Details
| Gene Symbol: Rapgef5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175930.5 | NP_787126.3 | 13 - 14 | 1635 | 85 | |
| GenBank mRNA | AK029995.1 | - | 13 - 14 | 1636 | 85 | |
| AK030016.1 | - | 13 - 14 | 1589 | 85 | ||
| AK043108.1 | - | 1 - 2 | 278 | 85 | ||
| AK083591.1 | - | 3 - 4 | 575 | 85 | ||
| AK122234.1 | - | 1 - 2 | 397 | 85 | ||
| BC046627.1 | - | 1 - 2 | 356 | 6494175 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rapgef5
Genomic Map