Assay Details
| Gene Symbol: Rapgef5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175930.5 | NP_787126.3 | 16 - 17 | 1961 | 142 | |
| GenBank mRNA | AK029995.1 | - | 16 - 17 | 1962 | 142 | |
| AK030016.1 | - | 16 - 17 | 1915 | 142 | ||
| AK083591.1 | - | 6 - 7 | 901 | 142 | ||
| AK122234.1 | - | 4 - 5 | 723 | 142 | ||
| BC046627.1 | - | 4 - 5 | 682 | 6494175 | 142 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rapgef5
Genomic Map