Assay Details
| Gene Symbol: G6pc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175935.3 | NP_787949.2 | 3 - 4 | 643 | 65 | |
| GenBank mRNA | AK002966.1 | - | 4 - 5 | 506 | 65 | |
| AK165395.1 | - | 3 - 4 | 65 | |||
| AY186239.1 | - | 3 - 4 | 489 | 65 | ||
| BC069959.1 | - | 3 - 4 | 643 | 4990136 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

G6pc3
Genomic Map