Assay Details
| Gene Symbol: Btn2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289615.1 | NP_001276544.1 | 5 - 6 | 1094 | 101 | |
| NM_175938.3 | NP_787952.2 | 5 - 6 | 1153 | 101 | ||
| GenBank mRNA | AK051061.1 | - | 5 - 6 | 1140 | 101 | |
| AY177686.1 | - | 5 - 6 | 1136 | 101 | ||
| BC139760.1 | - | 5 - 6 | 980 | 40058494 | 101 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of phosphatidylinositol 3-kinase signaling
negative regulation of cellular metabolic process
positive regulation of regulatory T cell differentiation
negative regulation of activated T cell proliferation
negative regulation of cytokine secretion
negative regulation of T cell receptor signaling pathway
negative regulation of protein kinase B signaling
negative regulation of ERK1 and ERK2 cascade
negative regulation of G1/S transition of mitotic cell cycle
molecular_function
negative regulation of phosphatidylinositol 3-kinase signaling
negative regulation of cellular metabolic process
positive regulation of regulatory T cell differentiation
negative regulation of activated T cell proliferation
negative regulation of cytokine secretion
negative regulation of T cell receptor signaling pathway
negative regulation of protein kinase B signaling
negative regulation of ERK1 and ERK2 cascade
negative regulation of G1/S transition of mitotic cell cycle
molecular_function
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2172540 | AY177686.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2172540 | BC139760.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2172540 | BC141341.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2172540 | NM_175938.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2172545 | NM_175938.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 677344 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 171195 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510767 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 522448 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 41199 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 131806 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107231 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165112 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2172545 | NM_175938.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2172540 | AY177686.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2172540 | BC139760.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2172540 | BC141341.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2172540 | NM_175938.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881734 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881742 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28881761 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107231 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165112 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07809801 | - | - |
| Whole Mouse Genome G4122A | A_51_P267913 | AK051061.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122552 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133637 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107231 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165112 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10839122 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10602137 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10839119 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10858328 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10983228 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10602136 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10858325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28881675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10850307 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11101363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881791 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881747 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10602137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881757 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11018399 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10874581 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881774 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10475813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881714 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881720 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11007307 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11018398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881734 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881759 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11104928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881779 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881755 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881743 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881773 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10826602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881790 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11007308 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881716 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881739 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881783 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881718 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10945254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881762 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10850305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881772 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10839122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11077010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881710 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881728 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881746 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881724 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10858328 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10651755 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28881721 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10408189 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408192 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408186 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408193 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408191 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408190 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408195 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408187 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408188 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408196 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408194 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408185 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 797770 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 520184 | AK018059.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 520184 | AK052578.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 520184 | BC004735.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 520184 | BC032218.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 520184 | NM_152822.2 | refseq_mrna_mouse_NM |
| Illumina mouse-6 v1.0 expression beadchip | 3390102 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003390102 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000520184 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1216723 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1229875 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1229875 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1216723 | - | - |
| mouse-6 v1.1 (Illumina) | scl0003714.1_245-S | - | - |
| mouse-6 v1.1 (Illumina) | scl44215.8_45-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3390102 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 520184 | AK018059.1 | genbank_mrna_mouse |
| Illumina Sentrix mouse-6.v1 bead array | 520184 | AK052578.1 | genbank_mrna_mouse |
| Illumina Sentrix mouse-6.v1 bead array | 520184 | BC004735.1 | genbank_mrna_mouse |
| Illumina Sentrix mouse-6.v1 bead array | 520184 | BC032218.1 | genbank_mrna_mouse |
| Illumina Sentrix mouse-6.v1 bead array | 520184 | NM_152822.2 | refseq_mrna_mouse_NM |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl44215.8_45-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003714.1_245-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 520184 | AK018059.1 | genbank_mrna_mouse |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 520184 | AK052578.1 | genbank_mrna_mouse |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 520184 | BC004735.1 | genbank_mrna_mouse |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 520184 | BC032218.1 | genbank_mrna_mouse |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 520184 | NM_152822.2 | refseq_mrna_mouse_NM |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3390102 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1216723 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2968694 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1216723 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2968694 | - | - |

Btn2a2
Genomic Map