Assay Details
| Gene Symbol: Osbpl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176840.3 | NP_789810.2 | 11 - 12 | 2791 | 90 | |
| GenBank mRNA | AK035188.1 | - | 1 - 2 | 174 | 90 | |
| BC035278.1 | - | 11 - 12 | 2791 | 5043236 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Osbpl11
Genomic Map