Assay Details
| Gene Symbol: Ddhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039106.3 | NP_001034195.2 | 3 - 4 | 1231 | 87 | |
| NM_001042719.2 | NP_001036184.1 | 4 - 5 | 1333 | 87 | ||
| NM_001284399.1 | NP_001271328.1 | 5 - 6 | 1305 | 87 | ||
| NM_176845.5 | NP_789815.3 | 3 - 4 | 1231 | 87 | ||
| GenBank mRNA | AK014975.1 | - | 1 - 2 | 81 | 87 | |
| AK048690.1 | - | 4 - 5 | 791 | 87 | ||
| AK129425.1 | - | 1 - 2 | 642 | 87 | ||
| AK155294.1 | - | 5 - 6 | 1262 | 87 | ||
| AK170271.1 | - | 4 - 5 | 945 | 87 | ||
| BC043475.1 | - | 2 - 3 | 485 | 5365658 | 87 | |
| CF532335.1 | - | 2 - 3 | 153 | 87 | ||
| DQ272482.1 | - | 3 - 4 | 1234 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddhd1
Genomic Map