Assay Details
| Gene Symbol: Zfp653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177318.2 | NP_796292.1 | 1 - 2 | 313 | 97 | |
| GenBank mRNA | AK046492.1 | - | 1 - 2 | 235 | 97 | |
| AK089104.1 | - | 1 - 2 | 125 | 97 | ||
| AK157635.1 | - | 1 - 2 | 235 | 97 | ||
| AK171735.1 | - | 1 - 2 | 335 | 97 | ||
| AY072705.1 | - | 1 - 2 | 299 | 97 | ||
| BC038037.1 | - | 1 - 2 | 313 | 4190337 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp653
Genomic Map