Assay Details
| Gene Symbol: Zfp653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177318.2 | NP_796292.1 | 3 - 4 | 568 | 85 | |
| GenBank mRNA | AK046492.1 | - | 3 - 4 | 490 | 85 | |
| AK089104.1 | - | 3 - 4 | 380 | 85 | ||
| AK157635.1 | - | 3 - 4 | 490 | 85 | ||
| AY072705.1 | - | 3 - 4 | 554 | 85 | ||
| BC038037.1 | - | 3 - 4 | 568 | 4190337 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26350331 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26350324 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26350250 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26350251 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26350330 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26350332 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26350334 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2035023 | BC038037.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2035023 | AK046492.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2035023 | NM_177318.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 298916 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 253499 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 162217 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137120 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 65511 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 43316 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 63857 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 964106 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 104950 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169717 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173182 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109650 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2035023 | BC038037.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2035023 | AK046492.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2035023 | NM_177318.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P398141 | AY072705.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P398141 | BC038037.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P398141 | AK046492.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P398141 | AK089104.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P398141 | AK157635.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350320 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350322 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26350249 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173182 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169717 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109650 | - | - |
| Whole Mouse Genome G4122A | A_51_P398141 | AY072705.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P398141 | BC038037.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P398141 | AK046492.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P398141 | AK089104.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P398141 | AK157635.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134687 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138925 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173182 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109650 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169717 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08111535 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08073242 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26350331 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26350324 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26350250 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26350332 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26350251 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26350330 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26350334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26350302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07823665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08065930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07703507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07464546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08193177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07823669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07823684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08193179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07823664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07708241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08168102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07893211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07461923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07784339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07464544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350318 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08036481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08057859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26350311 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10591718 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591724 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591719 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591716 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591725 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591723 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591722 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591717 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591720 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591721 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435481_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10591715 | - | - |
| Mouse Genome 430 2.0 Array | 1435481_at | - | - |
| Mouse Genome 430 2.0 Array | 1455560_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435481_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455560_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 319601_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 319601_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900015988.mm.1 | NM_177318.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900015988.mm.1 | TC0900001895.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1435481_PM_at | - | - |
| Mouse Expression Set 430 | 1435481_at | - | - |
| Murine Genome U74v2 Set | 104666_PM_at | - | - |
| Murine Genome U74v2 Set | 104666_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435481_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435481_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 724265 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 610717 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 610717 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000610717 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2755621 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2755621 | - | - |
| mouse-6 v1.1 (Illumina) | scl0319601.1_34-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 610717 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0319601.1_34-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 610717 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2755621 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2755621 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0319601.1_34-S | - | - |

Zfp653
Genomic Map