Assay Details
| Gene Symbol: Zfp653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177318.2 | NP_796292.1 | 4 - 5 | 1180 | 69 | |
| GenBank mRNA | AK089104.1 | - | 4 - 5 | 992 | 69 | |
| BC038037.1 | - | 4 - 5 | 1180 | 4190337 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp653
Genomic Map