Assay Details
| Gene Symbol: Myo1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177390.3 | NP_796364.2 | 12 - 13 | 1768 | 113 | |
| GenBank mRNA | AK049374.1 | - | 12 - 13 | 1769 | 113 | |
| BC150765.1 | - | 12 - 13 | 1634 | 9087676 | 113 | |
More Information
Set Membership: |
|

Myo1d
Genomic Map