Assay Details
| Gene Symbol: Myo1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177390.3 | NP_796364.2 | 16 - 17 | 2354 | 78 | |
| GenBank mRNA | AK049374.1 | - | 16 - 17 | 2355 | 78 | |
| AK148112.1 | - | 3 - 4 | 473 | 78 | ||
| BC034071.1 | - | 2 - 3 | 334 | 78 | ||
| BC039700.1 | - | 11 - 12 | 1486 | 78 | ||
| BC150765.1 | - | 16 - 17 | 2220 | 9087676 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Myo1d
Genomic Map