Assay Details
| Gene Symbol: Sun3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290519.1 | NP_001277448.1 | 4 - 5 | 243 | 109 | |
| NM_001290520.1 | NP_001277449.1 | 2 - 3 | 166 | 109 | ||
| NM_177576.3 | NP_808244.1 | 3 - 4 | 270 | 109 | ||
| GenBank mRNA | AK052771.1 | - | 3 - 4 | 271 | 109 | |
| AK132922.1 | - | 4 - 5 | 244 | 109 | ||
| BC109334.2 | - | 2 - 3 | 80 | 40051782 | 109 | |
| HQ141857.1 | - | 4 - 5 | 233 | 109 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK052771.1, BC109334.2, NM_001290520.1, NM_177576.3 transcripts.
Set Membership: |
|

Sun3
Genomic Map