Assay Details
| Gene Symbol: Tmem164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199360.1 | NP_001186289.1 | 1 - 2 | 208 | 84 | |
| GenBank mRNA | AK089342.1 | - | 1 - 2 | 58 | 84 | |
| BB842884.1 | - | 1 - 2 | 208 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK089342.1, NM_001199360.1 transcripts.
Set Membership: |
|

Tmem164
Genomic Map